ESD safety shoes, lace-up, CF 2
Supplier: ATLAS - the shoe company
76539 76540 76541 76542 76543 76544 76545 76546 76547 76549 76536 76537 76548 76538
114-1478EA 0 EUR
114-1478 114-1479 114-1480 114-1481 114-1482 114-1483 114-1484 114-1485 114-1486 114-1488 114-1475 114-1476 114-1487 114-1477
ESD safety shoes, lace-up, CF 2
Shoes and Boots
ESD safety shoe with safety reflectors and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
  • Carbon fibre reinforced toe protection
  • Electrical contact resistance of 1×10⁵ - 3,5×10⁷ Ohm
Colour: Black/blue
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1