Safety shoes, Velcro® fastening, TX 360
Supplier: ATLAS - the shoe company
83039 83040 83041 83042 83043 83044 83045 83046 83047 83038 83037 83036 83048 83049 83036/W12 83037/W12 83038/W12 83039/W12 83040/W12 83041/W12 83042/W12 83043/W12 83044/W12 83045/W12 83046/W12 83047/W12 83048/W12 83049/W12
114-1573EA 205 EUR
114-1573 114-1575 114-1577 114-1579 114-1581 114-1583 114-1585 114-1587 114-1588 114-1571 114-1569 114-1567 114-1589 114-1590 114-1568 114-1570 114-1572 114-1574 114-1576 114-1578 114-1580 114-1582 114-1584 114-1586 114-2750 114-2762 114-2763 114-2764
Safety shoes, Velcro® fastening, TX 360
Shoes and Boots
Lightweight safety shoe made from perforated Sportline upper (oiled nubuck leather), breathable functional lining and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: Pressure-relieving insole, cushioning comfort midsole made from MPU, dynamic outsole system
  • Protective steel toecap (200 J)
  • Carbon fibre reinforced toe protection