Glass fibre mats
Supplier: VOLZ LUFTFILTER
PSGRUN4110 PSGRUN4_125 MRGRUN420163 MRGRUN22050 MRGRUN22075 MRGRUN220100 MRGRUN220150 MRGRUN225100 MRGRUN32050 MRGRUN32075 MRGRUN320100 MRGRUN32550 MRGRUN32575 MRGRIN325100 MRGRIN325150 MRGRIN42050 MRGRIN42075 MRGRIN420100 MRGRIN420150 MRGRIN425100 MRGRIN425150 MRGRIN320200
VOLZPSGRUN4110EA 0 EUR
VOLZPSGRUN4110 VOLZPSGRUN4_125 VOLZMRGRUN420163 VOLZMRGRUN22050 VOLZMRGRUN22075 VOLZMRGRUN220100 VOLZMRGRUN220150 VOLZMRGRUN225100 VOLZMRGRUN32050 VOLZMRGRUN32075 VOLZMRGRUN320100 VOLZMRGRUN32550 VOLZMRGRUN32575 VOLZMRGRIN325100 VOLZMRGRIN325150 VOLZMRGRIN42050 VOLZMRGRIN42075 VOLZMRGRIN420100 VOLZMRGRIN420150 VOLZMRGRIN425100 VOLZMRGRIN425150 VOLZMRGRIN320200
Glass fibre mats
Filters Glass Fibre Filters
Paint-Stop (green): For use in separation of ink mist in paint shops and colour spraying cabins. Can be used in the automotive industry, manufacturing plants for furniture, windows, machines etc. Contains no silicone or other lacquer harming substances. Fire prevention requirements according to DIN 53438 (F3).
Dust-Stop (yellow): For separation of dry dust types. Can be used in carpenters' workshops, metal working industry etc. Contains no silicone. Fire prevention requirements according to DIN 53438 (F3). Additionally impregnated with antibacterial dust adhesive.