Gloves, vinyl, Nitras® vinyl P / vinyl PF
Supplier: AS ARBEITSSCHUTZ
8101S 8101M 8101L 8101XL
ASAS8101SEA 0 EUR
ASAS8101S ASAS8101M ASAS8101L ASAS8101XL
Gloves, vinyl, Nitras® vinyl P / vinyl PF
Gloves
100% vinyl disposable gloves.
  • Available powder-free or powdered
  • With rolled edge
  • Packed in a practical dispenser box
Certifications: EN455; AQL 1.5
Approved for food contact