Supplier: KIND ARBEITSSICHERHEIT
82436M 82436L 824361XL 824362XL
113-08372EA 0 EUR
113-08372 113-08371 113-08369 113-08370
Workwear
Multifunctional jacket made of Gammatex®, with weather and flame protection according to the latest standards. The fabric consists of 98% polyester and 2% carbon fibers. The coating is 100% polyurethane, hydrophilic and breathable. The COTTEX® lining has the highest flame retardant index 3.
  • Flame retardant, anti-static, chemical proof, waterproof and breathable
  • Removable flame retardant body and sleeve thermal lining with inner pocket
  • Hood (adjustable) with drawstring and peak in the collar
  • Two-way front zip to collar end, Velcro cover
  • Inside cuffs, knitted waistband as a windbreak, Velcro straps for width adjustment
  • Two chest and side pockets, one zip pocket
Certifications: EN 340, EN ISO 14116, EN 1149-3, EN 13034, EN 343 3/2; Kat. III