N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14)
Supplier: G Bioscienes
Synonyms: 3-(N,N-Dimethylmyristylammonio)propanesulphonate, Tetradecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner Salt
DG058 DG059
DG058EA 135 EUR
DG058 DG059
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14)
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-14, SB-14)
Formula: C₁₉H₄₁NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00036910
CAS Number: 14933-09-6
EINECS: 239-003-9