AMPSO

Supplier: Roth Carl

Synonyms: N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulphonic acid

7159.1 7159.2
ROTH7159.1EA 0 EUR
ROTH7159.1 ROTH7159.2
AMPSO
AMPSO
Formula: C₇H₁₇NO₅S
Storage Temperature: Ambient
MDL Number: MFCD00041777
CAS Number: 68399-79-1
EINECS: 269-991-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR