Certified colour reference standards for spectrophotometers
Supplier: Thermo Fisher Scientific
9423UV95520E
9423UV95100E
401322989841
SPET9423UV95520EEA
2360
EUR
SPET9423UV95520E
SPET9423UV95100E
SPET401322989841
Certified colour reference standards for spectrophotometers
Standards
Colour Reference and Spectroscopy Standards
These colour reference standards can be used with UV-Vis spectroscopy.
- Each standard includes a calibration certificate
Certifications: Traceable to ISO/IEC Standard 17025 accredited calibration process.