Hydroxypropyl cellulose, M.W. 80000, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
435007-100G 435007-250G 435007-5G
SIAL435007-100GEA 118 EUR
SIAL435007-100G SIAL435007-250G SIAL435007-5G
Hydroxypropyl cellulose, M.W. 80000, Sigma-Aldrich®
Hydroxypropyl cellulose
MDL Number: MFCD00132688
CAS Number: 9004-64-2