Hydroxypropyl methylcellulose (Hypromellose), 15 mPa.s (2% in H2O), Sigma-Aldrich®
Supplier: Merck
Synonyms: Methocel K100, Hydroxypropyl methyl cellulose
M-Clarity™ quality level = MQ100
09963-100G 09963-25G
SIAL09963-100GEA 177 EUR
SIAL09963-100G SIAL09963-25G
Hydroxypropyl methylcellulose (Hypromellose), 15 mPa.s (2% in H2O), Sigma-Aldrich®
Hydroxypropyl methyl cellulose (Hypromellose)
Formula: C₃H₈O₂·xCH₄O
MDL Number: MFCD00131360
CAS Number: 9004-65-3