2-Deoxy-D-glucose, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

D6134-1G D6134-250MG D6134-25G D6134-5G
SIALD6134-1GEA 106 EUR
SIALD6134-1G SIALD6134-250MG SIALD6134-25G SIALD6134-5G
2-Deoxy-D-glucose, Sigma-Aldrich®
2-Deoxy-D-glucose
Formula: C₆H₁₂O₅
MDL Number: MFCD00151328
CAS Number: 154-17-6
EINECS: 205-823-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR