Ammonium iron (III) citrate, Reagent Grade
Supplier: VWR Chemicals
Synonyms: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate
0846-500GEA 239 EUR
0846-500G 0846-100G
Ammonium iron (III) citrate, Reagent Grade
Ammonium iron (III) citrate
Formula: C₆H₅+4y)Fe(x)N(y)O₇
Storage Temperature: Ambient
MDL Number: MFCD00013099
CAS Number: 1185-57-5
EINECS: 214-686-6
Order Now
Specification Test Results
Arsenic ≤0.0004%
Iron (III) 20.5 – 22.5%
Lead ≤0.003%
Solubility (5.0%, Water) Pass
Zinc ≤0.1%