m-Cresol purple, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2',2''-Dimethylphenolsulphonphthalein, m-Cresolsulphonphthalein, MCP, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

M-Clarity™ quality level = MQ200

857890-10G 857890-1G 857890-5G
SIAL857890-10GEA 73.9 EUR
SIAL857890-10G SIAL857890-1G SIAL857890-5G
m-Cresol purple, Sigma-Aldrich®
m-Cresol purple
Formula: C₂₁H₁₈O₅S
MW: 382.43 g/mol
MDL Number: MFCD00005871
CAS Number: 2303-01-7
EINECS: 218-960-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR