Cresol red, Sigma-Aldrich®
Supplier: Merck
Synonyms:
o-Cresolsulphonphthalein, 3,3-Bis(4-hydroxy-3-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
M-Clarity™ quality level = MQ200
114472-25G
114472-5G
SIAL114472-25GEA
67.9
EUR
SIAL114472-25G
SIAL114472-5G
Cresol red, Sigma-Aldrich®
Cresol red
Formula:
C₂₁H₁₈O₅S Storage Temperature: Ambient |
MDL Number:
MFCD00005878 CAS Number: 1733-12-6 EINECS: 217-064-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...