Biebrich scarlet, powder C.I. 26905
Supplier: MP Biomedicals
Synonyms: Bonceau BS, C.I. 26905, Acid Red 66, Double Scarlet BSF, Croceine Scarlet, Biebrich Scarlet sodium salt
0215485525 0215485580
ICNA0215485525EA 51.2 EUR
ICNA0215485525 ICNA0215485580
Biebrich scarlet, powder C.I. 26905
Biebrich Scarlet
Soluble in water (0,1% - clear red solution) and ethanol (1 mg/ml).
Dye content: ~ 60% (Lit.)
Soluble in water (0,1% - clear red solution) and ethanol (1 mg/ml).
Formula: C₂₂H₁₄N₄Na₂O₇S₂
MW: 556.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003891
CAS Number: 4196-99-0
EINECS: 224-084-5