Diphenylphosphinic acid

Supplier: Cayman Chemical

10011337-10 10011337-100 10011337-50 10011337-250
CAYM10011337-10EA 76 EUR
CAYM10011337-10 CAYM10011337-100 CAYM10011337-50 CAYM10011337-250
Diphenylphosphinic acid
Diphenylphosphinic acid
Formula: C₁₂H₁₁O₂P
MDL Number: MFCD00002132
CAS Number: 1707-03-5
EINECS: 216-948-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR