Carbenicillin disodium salt, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na
M-Clarity™ quality level = MQ200
C3416-10G C3416-1G C3416-250MG C3416-5G
SIALC3416-10GEA 1000 EUR
SIALC3416-10G SIALC3416-1G SIALC3416-250MG SIALC3416-5G
Carbenicillin disodium salt, Sigma-Aldrich®
Carbenicillin disodium salt
Formula: C₁₇H₁₆N₂O₆S·2Na
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6
EINECS: 225-360-8