Methyl green zinc chloride salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Warning
Synonyms: CI 42590, Ethyl Green, Methyl green zinc chloride double salt, Methyl Green
MQ200
67060-10G 67060-50G
67060-10GEA 159 EUR
67060-10G 67060-50G
Methyl green zinc chloride salt, Sigma-Aldrich®
Methyl Green zinc chloride salt
Formula: C₂₇H₃₅BrClN₃·xZnCl₂
MW: 653.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151094
CAS Number: 7114-03-6
EINECS: 230-415-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e323a451158bd057cfddc2efca3de6704f647323&vid=30840a8b2079b16f9e52713ad84a297949b0ce84&prid=web&clid=SAPDAM