(4-Nitrophenyl)-β-D-galactopyranoside hydrate ≥98%
Supplier: Thermo Fisher Scientific
Purity: 98+%
CAS No.: 200422-18-0
Molecular Formula: C12H15NO8.xH2O
Formula Weight: 319.27 (301.25anhy)
Storage Temperature: 2°C to 8°C
Melting Point: 180-184°
Beilstein Reference: 92213
Optical Rotation: -84° (c=0.3 in water)
MDL No.: MFCD00150725
Formula:
C₁₂H₁₅NO₈·H₂O Melting Pt: 180…184 °C |
MDL Number:
MFCD00150725 CAS Number: 200422-18-0 EINECS: 221-584-5 |