CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt), Sigma-Aldrich®

Supplier: Merck

Synonyms: Sodium 3-(cyclohexylamino)-2-hydroxypropane-1-sulphonate

M-Clarity™ quality level = MQ200

C2154-100G C2154-1KG
SIALC2154-100GEA 137 EUR
SIALC2154-100G SIALC2154-1KG
CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt), Sigma-Aldrich®
CAPSO sodium salt (3-(Cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt)
Formula: C₉H₁₈NNaO₄S
MDL Number: MFCD00070063
CAS Number: 102601-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR