2-Naphthyl phosphate monosodium salt, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
N7375-10G N7375-1G N7375-5G
SIALN7375-10GEA 630 EUR
SIALN7375-10G SIALN7375-1G SIALN7375-5G
2-Naphthyl phosphate monosodium salt, Sigma-Aldrich®
2-Naphthyl phosphate monosodium salt
Formula: C₁₀H₈NaO₄P
MDL Number: MFCD00065914
CAS Number: 14463-68-4
EINECS: 238-452-8