Niobium(V) ethanolate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Niobium(V) ethoxide
M-Clarity™ quality level = MQ100
339202-50G
339202-5G
SIAL339202-50GEA
616
EUR
SIAL339202-50G
SIAL339202-5G
Niobium(V) ethanolate, Sigma-Aldrich®
Niobium(V) ethanolate
Formula:
C₁₀H₂₅NbO₅ MW: 318.209 g/mol |
MDL Number:
MFCD00015122 CAS Number: 3236-82-6 EINECS: 221-795-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...