Chromotrope 2R, pure
Supplier: Thermo Fisher Scientific
Warning
192010250 192011000
ACRO192010250EA 63.2 EUR
ACRO192010250 ACRO192011000
Chromotrope 2R, pure
Chromotrope 2R
Formula: C₁₆H₁₀N₂Na₂O₈S₂
MW: 468.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003955
CAS Number: 4197-07-3
EINECS: 224-085-0
Order Now
Specification Test Results
Appearance Red-brown Crystalline powder
Infrared spectrum Conforms
Specific extinction E (1%/1cm) ≥360 (509 nm, water)