Chromotrope 2R
Supplier: Thermo Fisher Scientific
A17519.14 A17519.22
A17519.14EA 50.6 EUR
A17519.14 A17519.22
Chromotrope 2R
Chromotrope 2R
CAS No.: 4197-07-3 Molecular Formula: C16H10N2Na2O8S2 Formula Weight: 468.37 MDL No.: MFCD00003955
Formula: C₁₆H₁₀N₂Na₂O₈S₂
MW: 468.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003955
CAS Number: 4197-07-3
EINECS: 224-085-0