Tobramycin, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol

M-Clarity™ quality level = MQ200

T4014-100MG T4014-500MG
SIALT4014-100MGEA 220 EUR
SIALT4014-100MG SIALT4014-500MG
Tobramycin, Sigma-Aldrich®
Tobramycin
Formula: C₁₈H₃₇N₅O₉
Storage Temperature: Refrigerator
MDL Number: MFCD00077885
CAS Number: 32986-56-4
EINECS: 251-322-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR