3'-Deoxyadenosine (cordycepine), Sigma-Aldrich®
Supplier: Merck
Synonyms: 3'-Deoxyadenosine, (2R,3R,5S)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3-ol, Cordycepin
C3394-100MG C3394-10MG C3394-25MG
SIALC3394-100MGEA 828 EUR
SIALC3394-100MG SIALC3394-10MG SIALC3394-25MG
3'-Deoxyadenosine (cordycepine), Sigma-Aldrich®
3'-Deoxyadenosine (Cordycepine)
Formula: C₁₀H₁₃N₅O₃
MW: 251.24 g/mol
MDL Number: MFCD00037998
CAS Number: 73-03-0
EINECS: 200-791-4
UN: 2811
ADR: 6.1 III