Adenosine 5'-diphosphate disodium salt (ADP disodium salt), Sigma-Aldrich®
Supplier: Merck
Synonyms: Adenosine 5'-Diphosphate Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate
M-Clarity™ quality level = MQ100
01897-1G 01897-250MG
SIAL01897-1GEA 112 EUR
SIAL01897-1G SIAL01897-250MG
Adenosine 5'-diphosphate disodium salt (ADP disodium salt), Sigma-Aldrich®
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Formula: C₁₀H₁₃N₅Na₂O₁₀P₂
MW: 471.167 g/mol
MDL Number: MFCD00066635
CAS Number: 16178-48-6
EINECS: 240-314-7