Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate
M-Clarity™ quality level = MQ200
A3377-5G A3377-500MG A3377-100G A3377-10G A3377-1G A3377-25G A3377-500G
SIALA3377-5GEA 109 EUR
SIALA3377-5G SIALA3377-500MG SIALA3377-100G SIALA3377-10G SIALA3377-1G SIALA3377-25G SIALA3377-500G
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) hydrate
Formula: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MW: 551.14 g/mol (anhy)
MDL Number: MFCD00150755
CAS Number: 34369-07-8
EINECS: 213-579-1