Actinomycin D ~98% (by HPLC), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
A1410-5MG A1410-100MG A1410-10MG A1410-25MG A1410-2MG
SIALA1410-5MGEA 249 EUR
SIALA1410-5MG SIALA1410-100MG SIALA1410-10MG SIALA1410-25MG SIALA1410-2MG
Actinomycin D ~98% (by HPLC), Sigma-Aldrich®
Actinomycin D
Formula: C₆₂H₈₆N₁₂O₁₆
MDL Number: MFCD00005033
CAS Number: 50-76-0
UN: 3462
ADR: 6.1,II