Actinomycin D ~95% (by HPLC), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
A4262-2MG
A4262-10MG
A4262-25MG
A4262-5MG
SIALA4262-2MGEA
83
EUR
SIALA4262-2MG
SIALA4262-10MG
SIALA4262-25MG
SIALA4262-5MG
Actinomycin D ~95% (by HPLC), Sigma-Aldrich®
Actinomycin D
Formula:
C₆₂H₈₆N₁₂O₁₆ |
MDL Number:
MFCD00005033 CAS Number: 50-76-0 UN: 3462 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...