Octyl-β-D-thioglucopyranoside, Sigma-Aldrich®
Supplier: Merck
Synonyms: Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside
MQ200
O6004-10G O6004-1G O6004-5G
SIALO6004-10GEA 676 EUR
SIALO6004-10G SIALO6004-1G SIALO6004-5G
Octyl-β-D-thioglucopyranoside, Sigma-Aldrich®
Octyl β-D-thioglucopyranoside
Formula: C₁₄H₂₈O₅S
MW: 308.44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012189
CAS Number: 85618-21-9