(1S)-(-)-Camphanic chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: (1S)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride, (-)-Camphanic acid chloride

M-Clarity™ quality level = MQ200

226173-1G 226173-5G
SIAL226173-1GEA 42.3 EUR
SIAL226173-1G SIAL226173-5G
(1S)-(-)-Camphanic chloride, Sigma-Aldrich®
(1S)-(-)-Camphanic chloride
Formula: C₁₀H₁₃ClO₃
MW: 216.67 g/mol
MDL Number: MFCD00135626
CAS Number: 39637-74-6
EINECS: 254-552-4
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR