Acridine orange (hemizinc salt), Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

A6014-100G A6014-10G A6014-25G
SIALA6014-100GEA 539 EUR
SIALA6014-100G SIALA6014-10G SIALA6014-25G
Acridine orange (hemizinc salt), Sigma-Aldrich®
Acridine orange (hemizinc salt)
Formula: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
Storage Temperature: Ambient
MDL Number: MFCD00081043
CAS Number: 10127-02-3
EINECS: 233-353-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR