Swab in tubes
Supplier: LP ITALIANA
111298 111598 111698
LPIT111298EA 300 EUR
LPIT111298 LPIT111598 LPIT111698
Swab in tubes
Swabs Swabs in Tubes
Sterile dry swabs in tubes.
Head is made of rayon or cotton.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904