Racks for pipette boxes
Supplier: GLW
P04 P06
GLWAP04EA 200 EUR
GLWAP04 GLWAP06
Racks for pipette boxes
Pipette Stands
These stainless steel racks are suitable for the storage of four or six GLW pipette boxes (A30, A40, A45 / S30, S40, S45).
Ordering information: Pipette boxes are not included.