Pycnometers, unadjusted
Supplier: witeg Labortechnik
3.900.025 3.900.050 3.900.001 3.900.002 3.900.005
613-2176EA 21.3 EUR
613-2176 613-2177 WITG3.900.001 WITG3.900.002 WITG3.900.005
Pycnometers, unadjusted
Specific Gravity Bottles
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in capillary stopper
  • Nominal capacity printed on the bottom
  • Very good chemical and temperature resistance
Certifications: DIN 12797