Anchorage lines, MH00
Supplier: HONEYWELL SAFETY
1002891 1002892 1002893 1002895
118-0421EA 90.3 EUR
118-0421 118-0422 118-0423 118-0425
Anchorage lines, MH00
Fall Protection Anchorage Lines
Twisted polyamide rope suitable for 14 mm/16 mm rope grabs. Suitable for vertical, horizontal and sloping applications.
  • Reliable and lightweight
  • Breaking strength: >22 KN
Certifications: EN 353-2: Guide type fall arresters on a flexible anchorage line Comply with the standard if used with MILLER rope grab only; EN 362 (rope)
Delivery information: Supplied with a counterweight and screwgate karabiner ML00 complies with EN696