Oxalyl dichloride 2 mol/l in dichloromethane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Oxalyl chloride
M-Clarity™ quality level = MQ200
310670-100ML 310670-800ML
SIAL310670-100MLEA 73.5 EUR
SIAL310670-100ML SIAL310670-800ML
Oxalyl dichloride 2 mol/l in dichloromethane, Sigma-Aldrich®
Oxalyl dichloride
Formula: C₂Cl₂O₂
MW: 126.93 g/mol
Boiling Pt: 63
Melting Pt: -12
Storage Temperature: Ambient
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
Merck Index: 12,07046