3,3-Dimethylbutyraldehyde ≥97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 3,3-Dimethylbutanal
H55522.AA H55522.XF
H55522.AAEA 265 EUR
H55522.AA H55522.XF
3,3-Dimethylbutyraldehyde ≥97%
3,3-Dimethylbutyraldehyde
Formula: C₆H₁₂O
Boiling Pt: 104…106 °C
Density: 0.798
Flash Pt: 13 °C (55 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042807
CAS Number: 2987-16-8
EINECS: 221-054-3
UN: 1989
ADR: 3,III