Ethyl-1,3-dithiane-2-carboxylate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 98+%
CAS No.: 20462-00-4
Molecular Formula: C7H12O2S2
Formula Weight: 192.30
Flash Point: 54°(129°F)
Refractive Index: 1.5400
Melting Point: 19-21°
Boiling Point: 75-77°/0.2mm
Beilstein Reference: 1424352
UN No.: 1993
MDL No.: MFCD00006657
Formula:
C₇H₁₂O₂S₂ Boiling Pt: 75…77 °C (0.2 mmHg) Melting Pt: 19…21 °C Density: 1.220 Flash Pt: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006657 CAS Number: 20462-00-4 EINECS: 243-838-4 ADR: 3,III |