Alcian blue 8GX Microscopy, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Warning
MQ100
05500-25G 05500-5G
05500-25GEA 683 EUR
05500-25G 05500-5G
Alcian blue 8GX Microscopy, Sigma-Aldrich®
Alcian blue 8 GX
Alcian Blue 8GX is used as a heteroglycan stain for neutral, sulfated and phosphated mucopolysaccharides and glycosaminoglycans in tissues such as cartilage and extracellular matrices.
Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
MW: 1298.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010720
CAS Number: 33864-99-2
EINECS: 251-705-7