Alcian blue 8GX, certified, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Warning
M-Clarity™ quality level = MQ200
A3157-10G A3157-25G A3157-10G A3157-25G
A3157-10GEA 278 EUR
A3157-10G A3157-25G SIALA3157-10G SIALA3157-25G
Alcian blue 8GX, certified, Sigma-Aldrich®
Alcian blue 8 GX
Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
Storage Temperature: Ambient
MDL Number: MFCD00010720
CAS Number: 33864-99-2
EINECS: 251-705-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cfc83cff437f130d3cd8d94e8407473c290f71e&vid=e92caad89207484fd795ac310f64a0d9a1b7cc3f&prid=web&clid=SAPDAM