N,N-Diphenylcarbamoyl chloride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
B23116.14
B23116.22
B23116.14EA
54.5
EUR
B23116.14
B23116.22
N,N-Diphenylcarbamoyl chloride ≥98%
N,N-Diphenylcarbamoyl chloride
Purity: 98%
CAS No.: 83-01-2
Molecular Formula: C13H10ClNO
Formula Weight: 231.68
Odour: Irritating
Melting Point: 82-86°
Beilstein Reference: 515312
UN No.: 3261
MDL No.: MFCD00000633
Formula:
C₁₃H₁₀ClNO Melting Pt: 82…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000633 CAS Number: 83-01-2 EINECS: 201-450-2 UN: 3261 ADR: 8,II |