Carbohydrazide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

C11006-100G C11006-25G
SIALC11006-100GEA 124 EUR
SIALC11006-100G SIALC11006-25G
Carbohydrazide, Sigma-Aldrich®
Carbohydrazide
Formula: CH₆N₄O
MDL Number: MFCD00007591
CAS Number: 497-18-7
EINECS: 207-837-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR