Milk, non fat (skimmed milk), powder
Supplier: BIOMOL RESEARCH LABORATORIES
54650-1 54650-2 54650-3
BMOL54650-1EA 74 EUR
BMOL54650-1 BMOL54650-2 BMOL54650-3
Milk, non fat (skimmed milk), powder
Non-fat milk powder (skim milk)
Storage Temperature: Ambient
CAS Number: 999999-99-4
EINECS: 310-127-6