Propanephosphonic acid cyclic anhydride 50% in ethyl acetate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Propylphosphonic anhydride, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxotriphosphorinane-2,4,6-trioxide
M-Clarity™ quality level = MQ200
431303-10ML 431303-50ML 431303-500ML
SIAL431303-10MLEA 42.1 EUR
SIAL431303-10ML SIAL431303-50ML SIAL431303-500ML
Propanephosphonic acid cyclic anhydride 50% in ethyl acetate, Sigma-Aldrich®
Propanephosphonic acid cyclic anhydride
Formula: C₉H₂₁O₆P₃
MW: 318.19 g/mol
MDL Number: MFCD00006583
CAS Number: 68957-94-8
EINECS: 422-210-5
UN: 2924
ADR: 3 II