Indomethacin, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

I7378-100G I7378-10G I7378-25G I7378-5G
SIALI7378-100GEA 581 EUR
SIALI7378-100G SIALI7378-10G SIALI7378-25G SIALI7378-5G
Indomethacin, Sigma-Aldrich®
Indomethacin
Formula: C₁₉H₁₆ClNO₄
MW: 357.80 g/mol
MDL Number: MFCD00057095
CAS Number: 53-86-1
EINECS: 200-186-5
UN: 2811
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR