Pyridoxal-5'-phosphate monohydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Pyridoxal-5-phosphate hydrate, Pyridoxal 5-phosphate monohydrate, Pyridoxal 5'-phosphate monohydrate, (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydrate
82870-1G 82870-25G 82870-5G
SIAL82870-1GEA 43.9 EUR
SIAL82870-1G SIAL82870-25G SIAL82870-5G
Pyridoxal-5'-phosphate monohydrate, Sigma-Aldrich®
Pyridoxal 5'-phosphate monohydrate
Formula: C₈H₁₀NO₆P·1H₂O
MW: 265.17 g/mol (247.15 anhy)
Storage Temperature: Freezer
MDL Number: MFCD00149414
CAS Number: 41468-25-1