Luminol (3-Aminophthalhydrazide) 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
NSC 5064, Luminol, 5-Amino-2,3-Dihydro-1,4-Phthalazinedione, 5-Amino-2,3-dihydrophthalazine-1,4-dione
A14597.06
A14597.14
A14597.22
A14597.06EA
70.2
EUR
A14597.06
A14597.14
A14597.22
Luminol (3-Aminophthalhydrazide) 98%
Luminol (3-Aminophthalhydrazide)
Purity: 98%
CAS No.: 521-31-3
Molecular Formula: C8H7N3O2
Formula Weight: 177.16
Odour: Odourless
Melting Point: 325-330°
Beilstein Reference: 383929
Merck Reference: 14,5600
MDL No.: MFCD00006890
Formula:
C₈H₇N₃O₂ Melting Pt: 325…330 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006890 CAS Number: 521-31-3 EINECS: 208-309-4 Merck Index: 14,5600 |