(±)-3-Carene 90% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
3,7,7-Trimethylbicyclo[4.1.0]-3-heptene, 3,7,7-Trimethylbicyclo(4.1.0)hept-3-ene, 3,7,7-Trimethylbicyclo[4.1.0]hept-3-ene, 3-Carene, (±)-3,7,7-Trimethylbicyclo[4.1.0]-3-heptene
367960250
367960010
ACRO367960250EA
31.3
EUR
ACRO367960250
ACRO367960010
(±)-3-Carene 90% stabilised
(±)-3-Carene
Formula:
C₁₀H₁₆ Boiling Pt: 167…170 °C (1013 hPa) Density: 0.864 g/cm³ (20 °C) Flash Pt: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001315 CAS Number: 13466-78-9 EINECS: 236-719-3 UN: 1993 ADR: 3,III Merck Index: 12,01885 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4720 to 1.4740 (20°C, 589 nm) |
Density | (20°C) 0.862 to 0.865 g/ml |
GC | ≥88 % |
Stabilizer | 0.01 % alpha-Tocopherol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...